Order Entry
United States
ContactUsLinkComponent
Pierce™ Immobilized Papain (Agarose Resin), Thermo Scientific
Pierce™ Immobilized Papain (Agarose Resin), Thermo Scientific
Catalog # PI20341
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ Immobilized Papain (Agarose Resin), Thermo Scientific
Catalog # PI20341
Supplier:  Invitrogen
Supplier Number:  20341
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Immobilized Papain (agarose resin)
  • Size:
    5 mL
  • Storage Temperature:
    +4°C
  • Cat. No.:
    PI20341
  • Supplier no.:
    20341

Specifications

About this item

Thermo Scientific Pierce Immobilized Papain consists of a cysteine-endopeptidase (papain) that has been immobilized onto beaded agarose resin to enable generation and purification of Fab fragments from antibodies.

  • Papain can also be used to generate F(ab')2 fragments
  • Immobilized papain canbe substituted fro free papain because it eliminates autolysis and contamination of a sample with the protease
  • Allows control of the digestion by removing the pepsin or controlling the flow of sample over an immobilized papain column
  • Immobilized papain is more stable against heat-induced denaturation, resulting in longer maintenance of activity

Papain is a nonspecific, cysteine-endopeptidase isolated from papaya latex that is used in a wide variety of applications. When IgG molecules are incubated with papain in the presence of cysteine, one or more peptide bonds in the hinge region are split, producing three fragments of similar size: two Fab fragments and one Fc fragment. The Fc fragments can be separated from the Fab fragments using immobilized Protein A or by ion exchange chromatography.

Caution: For Research Use Only. Not for use in diagnostic procedures.