Pierce™ Recombinant C-Myc Peptide
PI20170
:
- Protein/Peptide Type:Recombinant
- Size:5 mg
- Storage Conditions:–20˚C
- Protein/Peptide Name:C-Myc Peptide
- Tested Applications:Immunoprecipitation
- Cat. No.:PI20170
- Supplier no.:20170
The c-Myc peptide may be used to elute c-Myc-tagged proteins from Thermo Scientific Anti-c-Myc Agarose, when the bound protein is sensitive to conditions of chemical elution, such as extremes in pH, or sodium thiocyanate.
- Competes out the binding of monoclonal anti-c-Myc or rabbit anti-c-Myc antibodies
- Synthetic peptide provided as a lyophilized powder
- Used with water to prepare stock solution
The peptide is useful for competing out anti-c-Myc antibody binding to c-Myc-tagged fusion proteins in immunoassays. The successful inhibition of antibody binding ascertains that the experimental observed binding is specific.
: For Research Use Only. Not for use in diagnostic procedures.