Pierce™ Buffer, Sodium Acetate (3 M), pH 5.2, Thermo Scientific
Catalog # PI17888
Supplier: Invitrogen
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Sodium Acetate Buffer Solution, 3.0 M
- Size:1 L
- Environmentally Preferable:
- Storage Temperature:20...25 °C
- Cat. No.:PI17888
- pH:5.2
- Supplier no.:17888
Specifications
About this item
Thermo Scientific Pierce Sodium Acetate Solution is a sterile, 3M solution of sodium acetate that is useful for basic methods of DNA precipitation, DNA purification and storage.
- Helps in maintaining constant pH level
Caution: For Research Use Only. Not for use in diagnostic procedures.