

Trifluoroacetic acid ≥99.8%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Catalog # BDH15311.100
Supplier: VWR International
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle, amber
- Size:1 L
- Cat. No.:BDH15311.100
- Supplier no.:BDH15311.100
Specifications
Specification Test Results
- Assay by Titration:Min. 99.8 %
- Color:Max. 10 APHA
- Transmittance (210 nm) 0.1% aq. soln:Min. 30 %
- Transmittance (220 nm) 0.1% aq. soln:Min. 60 %
- Transmittance (230 nm) 0.1% aq. soln:Min. 85 %
- Transmittance (240 nm) 0.1% aq. soln:Min. 95 %
- Transmittance (250 nm) 0.1% aq. soln:Min. 99 %
Specification Test Results
About this item
:
:114.02 g/mol
Merck Index:13,09754
Boiling Pt:71.8 °C (1013 hPa)
Melting Pt:–15 °C
Density:1.490 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-929-3
UN:2699
ADR:8,I
:GHS05
:GHS07
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169