ContactUsLinkComponent
Pasteurella Multocida Toxin, List Biological Laboratories
Pasteurella Multocida Toxin, List Biological Laboratories
Catalog # 102946-474
CAS Number:  
Pasteurella Multocida Toxin, List Biological Laboratories
Catalog # 102946-474
Supplier Number:  156
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Pasteurella Multocida Toxin
  • Size:
    50 µg
  • Cat. No.:
    102946-474
  • Supplier no.:
    156
Specifications
About this item
Pasteurella multocida toxin is produced by some strains of Pasteurella multocida, a small gram-negative coccobacillus that is pathogenic for many species of animals
Purified Pasteurella multocida toxin has a molecular weight of approximately 146,000 daltons, corresponding to 1,285 amino acids. This toxin can affect the signal transduction pathway of many cells by selective transient activation of a G-protein known as Gqα.