About this item
Staphylococcus aureus is a common commensal of the skin and mucosal membranes which causes significant illness worldwide through production of numerous Staphylococcal toxins. Food poisoning can result from consumption of foods containing preformed enterotoxins, SEA, SEB, SEC, SED and SEE resulting from Staphylococcal organisms. Other illnesses caused by this organism include toxic shock syndrome, scalded skin syndrome, pneumonia, and infective endocarditis. Staphylococcal toxins that act on cell membranes include alpha toxin, beta toxin, delta toxin, and Panton-Valentine leukocidin.
VWR® offers for sale two Staphylococcal toxins and one toxoid. The products are enterotoxin B (SEB), alpha toxin and SEB toxoid. More information about each Staphylococcal toxin we have available may be obtained through the linked Technical Information.