ContactUsLinkComponent
Calbiochem®, Phosphatase Inhibitor Cocktail Set II, EMD Millipore
Calbiochem®, Phosphatase Inhibitor Cocktail Set II, EMD Millipore
Catalog # 80501-130
Supplier:  MilliporeSigma
CAS Number:  
Calbiochem®, Phosphatase Inhibitor Cocktail Set II, EMD Millipore
Catalog # 80501-130
Supplier:  MilliporeSigma
Supplier Number:  524625-1SET
CAS Number:  
Specifications
  • Description:
    Phosphatase inhibitors
  • Size:
    1 set (5 x 1 mL)
  • Cat. No.:
    80501-130
  • Supplier no.:
    524625-1SET
Specifications
About this item
A cocktail of five phosphatase inhibitors for the inhibition of acid and alkaline phosphatases as well as protein tyrosine phosphatases (PTPs)
Suitable for use with cell lysates and tissue extracts, including samples containing detergents.