Order Entry
United States
ContactUsLinkComponent
Antigen Preserve Solution VII, Electron Microscopy Sciences
Antigen Preserve Solution VII, Electron Microscopy Sciences
Catalog # 102091-804
CAS Number:  
Antigen Preserve Solution VII, Electron Microscopy Sciences
Catalog # 102091-804
Supplier Number:  1227SK
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Antigen Preserve Solution VII
  • Size:
    1 l
  • Cat. No.:
    102091-804
  • Supplier no.:
    1227SK

Specifications

About this item