Order Entry
United States
ContactUsLinkComponent
Sodium hydrogen carbonate 5% in aqueous solution
Irritant
Sodium hydrogen carbonate 5% in aqueous solution
Catalog # 101410-796
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
Irritant
Sodium hydrogen carbonate 5% in aqueous solution
Catalog # 101410-796
Supplier Number:  21127
Minority or Woman-Owned Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    500 ml
  • Cat. No.:
    101410-796
  • Supplier no.:
    21127

Specifications

About this item

 :
 :84.01 g/mol
Boiling Pt:100 °C (212 °F)
Density:1.06 g/cm³ (8.846 lbs/gal.)
 :GHS07
CAS Number:144-55-8
Formula:NaHCO₃
MDL Number:MFCD00003528