Acetone fixative reagent, buffered
500 ml
Catalog # 101410-606
Supplier: Electron Microscopy Sciences
: 10018
Acetone fixative reagent, buffered
Catalog # 101410-606
Supplier: Electron Microscopy Sciences
CAS Number: 67-64-1
Jump to:
Product Details & Documents
Acetone
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- UN:1090
- ADR:3,II
Specifications
Specifications
- Size:500 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



