Order Entry
United States
ContactUsLinkComponent
Lissamine Fast Red staining solution
Lissamine Fast Red staining solution
Catalog # 101410-906
Minority or Woman-Owned Business Enterprise
CAS Number:  
Lissamine Fast Red staining solution
Catalog # 101410-906
Supplier Number:  26034-05
Minority or Woman-Owned Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    500 ml
  • Environmentally Preferable:
  • Cat. No.:
    101410-906
  • Supplier no.:
    26034-05

Specifications

About this item

Synonyms: acid violet 7, Lissamine Fast Red, 5-(Acetylamino)-3-[[4-(acetylamino) phenyl] azo]-4-hydroxy-2,7- naphthalenedisulphonic acid disodium salt, C.I 18055, 5-(Acetylamino)-3-[[4-(acetylamino) phenyl] azo]-4-hydroxy-2,7- naphthalenedisulfonic acid disodium salt, acid Naphthol Red 6B
 :
 :566.5 g/mol
 :GHS07
CAS Number:4321-69-1
Formula:C₂₀H₁₆N₄O₉S₂Na₂
MDL Number:MFCD00003948