Order Entry
United States
ContactUsLinkComponent
 
Irritant
Cresyl violet (acetate) 0.1% in aqueous solution stain for histology
Catalog # 101411-000
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
Irritant
Cresyl violet (acetate) 0.1% in aqueous solution stain for histology
Catalog # 101411-000
Supplier Number:  26089-01
Minority or Woman-Owned Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 ml
  • Cat. No.:
    101411-000
  • Supplier no.:
    26089-01

Specifications

About this item

 :
 :321.33 g/mol
 :GHS07
CAS Number:10510-54-0
Formula:C₁₈H₁₅N₃O₃
MDL Number:MFCD00013151