Taft’s Method for Nucleic Acid, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Taft’s Method for Nucleic Acid (1951), Stain DNA & RNA
Stain results - DNA, Deoxryibonucleic Acid: Blue Green, RNA, Ribonucleic Acid: Bright Red.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.