About this item
Methenamine Silver Method for Argentaffin Cells for staining of argentaffin granules of enterochromaffin cells, Results - Argentaffin Cells: Black, Coarse connective tissue of submucosa: Variable amount of blackening.
Clark, G.: Staining Procedures, Williams and Wilkins Co., Baltimore, 3rd Ed., c. 1973, p. 161.
Gomori, G., Arch. Path., 45:48, 1948.
Burtner, H.J. and Lillie, R.D., Stain Techn., 24:225-7, 1949.