About this item
Dane’s Method for Prekeratin, Keratin, and Mucin. Keratin stain method for the staining of keratin, prekeratin, acid mucopolysaccharides and nuclei. Stain results: Acid Mucopolysaccharides: Blue, Prekeratin and Keranin: Orange to Red, Nuclei: Brown - Orange
Dane, E.T., and Herman, D.L.: Stain Technology, 38:97, 1963.
AFIP Manual of Histological Staining Methods, 3rd Ed., Ed L. Luna: New York: McGraw Hill Publications, c 1968, p.83.
Dane’s Method for Prekeratin, Keratin, and Mucin
Keratin stain method for the staining of keratin, prekeratin, acid mucopolysaccharides and nuclei. Stain results: Acid Mucopolysaccharides: Blue, Prekeratin and Keranin: Orange to Red, Nuclei: Brown - Orange