About this item
Carstairs Method for Fibrin and Platelets is used for the staining of fibrin and platelets
Stain results - 48 hours or more: Fibrin: Bright Red, Platelets Gray blue to navy, Collagen: Bright Blue, Muscle: Red, Red Blood Cells: Clear Yellow.