Order Entry
United States
Orders LinkContactUsLinkComponent
Sym-Collidine Buffer Kit, Electron Microscopy Sciences

Sym-Collidine Buffer Kit, Electron Microscopy Sciences

Sym-Collidine Buffer Kit, Electron Microscopy Sciences
Certificates
Sym-Collidine Buffer Kit, Electron Microscopy Sciences
Sym-Collidine Buffer Kit, Electron Microscopy Sciences

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

A very stable buffer, used with Osmium tetroxide; provides excellent fixation, better stability and buffering capacity.

The pH can be adjusted by varying the amount of hydrochloric acid in the final volume of 200 ml.

Preparation Instructions: Pour 9 ml of 2 N HCl; add 5.34 ml of Sym-Collidine, mix well and add distilled water up to 200 ml for a pH 7.4 solution (Bennett and Luft., Biophys and Biochem. Cytol., 6, 113, 1959).

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 100503-712
  • 100503-714
  • CAS No.:
  • 108-75-8
  • 108-75-8
  • Storage Temperature:
  • 0...5 °C

Specifications

Product Family Options

Product Information

  • pH
    Size
    Availability
    Price
  • 100503-712
    Sym-Collidine Buffer Kit
    Sym-Collidine Buffer Kit
    7.4
    1 set
    Log in for availability
    Log in for availability
    Each
    Each - $71.22
    $71.22
    Each
       $71.22

    Supplier Number: 11500
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100503-712
    CAS No.
    108-75-8
    Storage Temperature
    0...5 °C
    Minority or Woman-Owned Business Enterprise
  • 100503-714
    Sym-Collidine Buffer
    Sym-Collidine Buffer
    7.4
    5 sets
    Log in for availability
    Log in for availability
    Each
    Each - $204.27
    $204.27
    Each
       $204.27

    Supplier Number: 11510
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100503-714
    CAS No.
    108-75-8
    Storage Temperature
    Minority or Woman-Owned Business Enterprise

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.