Order Entry
United States
ContactUsLinkComponent
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I), certified
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I), certified
Catalog # 100503-906
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I), certified
Catalog # 100503-906
Supplier Number:  15670
Minority or Woman-Owned Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    0.5 g
  • Cat. No.:
    100503-906
  • Supplier no.:
    15670

Specifications

Specification Test Results

  • Assay:
    90%

Specification Test Results

About this item

 :
 :389.381 g/mol
Melting Pt:>360 °C (>680 °F)
Storage Temperature:Refrigerator
CAS Number:3326-32-7
Formula:C₂₁H₁₁NO₅S
MDL Number:MFCD00005063