Orders LinkContactUsLinkComponent
Bismuth(III) carbonate basic, purified
25 g
  100503-692
Bismuth(III) carbonate basic, purified
  100503-692
 :  5892-10-4
Bismuth(III) carbonate basic, purified
Bismuth(III) carbonate basic, purified
25 g
 :  5892-10-4
$61.18
Each (25g)
Can be used for staining synapses (Pfenninger, K., Sandri, C., Albert, K. and Eugster, C.H., 1969). Contribution of the problem of structural organization of the presynaptic area (Brain Res., 12:10).
 :  Bismuth carbonate oxide, Dibismuth carbonate dioxide, Bismuth subcarbonate
  •  :5892-10-4
  •  :(BiO)₂CO₃
  •  :MFCD00010877
  •  :509.97 g/mol
  •  :6.86 g/cm³ (57.247 lbs/gal.)
  • Size:
    25 g