Order Entry
United States
ContactUsLinkComponent
Ammonium heptamolybdate tetrahydrate ≥99%
Irritant
Ammonium heptamolybdate tetrahydrate ≥99%
Catalog # 100503-650
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
Irritant
Ammonium heptamolybdate tetrahydrate ≥99%
Catalog # 100503-650
Supplier Number:  10500
Minority or Woman-Owned Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 g
  • Cat. No.:
    100503-650
  • Supplier no.:
    10500

Specifications

About this item

 :
 :1235.86 g/mol
Melting Pt:287 °C (549 °F)
Density:1.782 g/cm³ (14.871 lbs/gal.)
 :GHS07
CAS Number:12054-85-2
 :12027-67-7
Formula:(NH₄)₆Mo₇O₂₄·4H₂O
MDL Number:MFCD00167059