Order Entry
United States
Orders LinkContactUsLinkComponent
Ammonia solution 28% ACS

Ammonia solution 28% ACS

Corrosive
Irritant
Ammonia solution 28% ACS
Certificates
Ammonia solution 28% ACS

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

NH4OH F.W. 35.05 CAS 1336-21-6 Specific Gravity: 0.91 Assay 28-30%

Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:28 °C (82 °F)
  • Density:0.8321 g/cm³ (6.944 lbs/gal.)
  • UN:3318
  • ADR:2.2,I
Specifications for Product Family

Specification Test Results

  • Assay:
    28-30%
  • Carbon Dioxide:
    0.002%
  • Chloride:
    0.5 ppm
  • Heavy Metals:
    0.5 ppm
  • Iron:
    0.2 ppm
  • Phosphate:
    2 ppm
  • Residue After Ignition:
    0.002%
  • Specific Gravity:
    0.91
  • Sulfate:
    2 ppm
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle
    450 ml
    Log in for availability
    Log in for availability
    Each (450ml)
    Each (450ml) - $43.58
    $43.58
    Each (450ml)
       $43.58

    Supplier Number: 10600
  • Bottle
    4×450 ml
    Log in for availability
    Log in for availability
    Each (450ml)
    Each (450ml) - $153.36
    $153.36
    Each (450ml)
       $153.36

    Supplier Number: 10601

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.