About this item
Agarose I™ is standard melting/gelling agarose, suitable for routine nucleic acid analytical/preservative applications
- CAS No.= 9012-36-6
Agarose I™ has a low EEO for shorter electrophoresis runs without compromising resolution. Available in fine powder or tablet.