Thioglycollate Broth with Resazurin, Biomerieux
95060-842
:
- Description:Thioglycolate Broth with Resazurin
- Cat. No.:95060-842
- Supplier no.:44001
Mostly used for detection anaerobic bacteria.
- Detects anaerobic bacteria
- Also allows detection of aerobic bacteria
Mostly used for detection of anaerobic bacteria but also allows detection of aerobic bacteria recommended by USP/EP/JP for sterility testing of pharmaceutical articles.