BioBall® MultiShot, For Quantitative Quality Control Microorganisms, bioMerieux
Supplier: bioMerieux
Certificates
About this item
BioBall® mixed kits contain the most relevant pharmacopoeia strains required for quantitative microbiology
BioBall® contains a precise number of microorganisms in a water soluble ball, delivering unprecedented accuracy for Quantitative Microbiological Quality Control. These BioBall® Multishot 550 are mean of between 500 and 600 cfu with a standard deviation of ≤ 10% of the mean. Designed for re-hydration into 1.1 mL of MultiShot 550 Re-Hydration Fluid to provide 10 x 100 µL doses of 50 cfu. Ideal for pharmaceutical microbiology QC where multiple positive controls in the 10 to 100 cfu range are needed.
Related Information
Certificates
Search by Lot NumberBioBall Available Strains
Datasheet
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together