Order Entry
United States
ContactUsLinkComponent
 
Boardwalk® High-Density Waste Can Liners
  102865-926
undefined
Boardwalk® High-Density Waste Can Liners
  102865-926
 :  BWK243306

 

  • Capacity:
    16 gal.
  • Thickness:
    6 µm
  • Cat. No.:
    102865-926
  • W×L:
    24×33"
  • Supplier no.:
    BWK243306

 

 

Performance bottom star seal to prevent leakage.

  • Use for non-abrasive materials
  • Coreless rolls
  • Natural color

Made from high-quality raw materials, mineral reinforced to enhance strength and puncture-resistance.