Order Entry
United States
ContactUsLinkComponent
McIndoe Forceps, OR Grade, Sklar
McIndoe Forceps, OR Grade, Sklar
Catalog # 10194-460
Supplier:  Sklar
undefined
McIndoe Forceps, OR Grade, Sklar
Catalog # 10194-460
Supplier:  Sklar
Supplier Number:  19-1230

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Length:
    152 mm (6")
  • Material:
    Stainless steel
  • Cat. No.:
    10194-460
  • Jaw type:
    Serrated
  • Tip style:
    Straight
  • Supplier no.:
    19-1230

Specifications

About this item

McIndoe Forceps are used to hold tissue, remove debris from wounds, place and remove dressings and packing.

They are non-toothed, thumb forceps with a stop peg to ensure that when used for holding tissue the grip is not crushing.