Specifications
- Length:241 mm (9½")
- Material:Stainless steel
- Cat. No.:10194-212
- Jaw type:Serrated
- Tip style:Straight
- Supplier no.:90-7895
Specifications
About this item
Clemetson Uterine Forceps are non-ratcheted, finger ring grasping forceps used in dilation and evacuation procedures.
This product is straight with fenestrated, serrated, tear drop tips.