Order Entry
United States
ContactUsLinkComponent
San Jamar® Oceans™ Universal Liquid Soap Dispenser
San Jamar® Oceans™ Universal Liquid Soap Dispenser
Catalog # 102865-842
undefined
San Jamar® Oceans™ Universal Liquid Soap Dispenser
Catalog # 102865-842
Supplier Number:  SJMS890TBK

Specifications

  • Capacity:
    800 ml
  • Color:
    Black
  • Description:
    Ocean soap dispenser
  • Cat. No.:
    102865-842
  • W×D×H:
    4.5×4.375×10.5"
  • Supplier no.:
    SJMS890TBK

Specifications

About this item

Versatile dispenser may be used in conjunction with three separate delivery systems: bulk liquids and gels, bag-in-box cartridges or bag refills

For use with all 800-mL Bag-In-Box Refills. Liquid, manual dispenser type. Wall mounting.

Included 800-mL bottle accommodates bulk liquid soaps and gel sanitizers. Break-resistant plastic shell easily mounts to walls. Security lock utilizes a standard San Jamar® waffle key.