Anti-B3GAT1 Rabbit Polyclonal Antibody
10577-714
: Abnova
- Antibody Type:Primary
- Antigen Name:Beta-1,3-glucuronyltransferase 1
- Antigen Symbol:B3GAT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:LEU7|GLCATP|CD57|NK1|GLCUATP|NK-1|B3GAT1|HNK1
- Storage Buffer:In 1x PBS, pH 7.4
- Sequence:MPKRRDILAIVLIVLPWTLLITVWHQSTLAPLLAVHKDEGSDPRRETPPGADPREYCTSDRDIVEVVRTEYVYTRPPPWSDTLPTIHVVTPTYSRPVQKAELTRMANTLLHVPNLHWLVVEDAPRRTPLTARLLRDTGLNYTHLHVETPRNYKLRGDARDPRIPRGTMQRNLALRWLRETFPRNSSQPGVVYFADDDNTYSLELFEEMRSTRRVSVWPVAFVGGLRYEAPRVNGAGKVVGWKTVFDPHRPFAIDMAGFAVNLRLILQRSQAYFKLRGVKGGYQESSLLRELVTLNDLEPKAANCTKILVWHTRTEKPVLVNEGKKGFTDPSVEI
- Storage Temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:B3GAT1 (ABM86204.1, 1 a.a. ~ 334 a.a) full-length human protein.
- Cat. No.:10577-714
- Supplier no.:H00027087-D01P
Rabbit polyclonal antibody raised against a full-length human B3GAT1 protein.
Recommended Dilutons: Western Blot (1 ug/mL)
Type: Primary
Antigen: B3GAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat