ContactUsLinkComponent
Jansen Bayonet Dressing Forceps, OR Grade, Sklar
Jansen Bayonet Dressing Forceps, OR Grade, Sklar
Catalog # 10198-544
Supplier:  Sklar
undefined
Jansen Bayonet Dressing Forceps, OR Grade, Sklar
Catalog # 10198-544
Supplier:  Sklar
Supplier Number:  70-4385
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Length:
    216 mm (8½")
  • Material:
    Stainless steel
  • Cat. No.:
    10198-544
  • Jaw type:
    Serrated
  • Tip style:
    Straight
  • Supplier no.:
    70-4385
Specifications
About this item
Jansen Bayonet Dressing Forceps are thumb forceps used in nasal procedures for placing packing
The bayonet style handle and long jaws allow an unobstructed view when working in the nasal cavity. Serrated tips hold surgical materials.
They may be used in intracranial procedures.