Order Entry
United States
ContactUsLinkComponent
Windsoft® Center-Flow Perforated Towels
Windsoft® Center-Flow Perforated Towels
Catalog # 102866-242
undefined
Windsoft® Center-Flow Perforated Towels
Catalog # 102866-242
Supplier Number:  WIN1420B

Specifications

  • Ø:
    8.6"
  • L×W:
    13.5 x 8"
  • Cat. No.:
    102866-242
  • Roll length:
    660'
  • Supplier no.:
    WIN1420

Specifications

About this item

Towels dispense singly.

  • Paper material
  • Unscented

Absorbent two-ply. 8³/₅" Ø roll. 7⁹/₁₀×13¹/₂ sheets. Towels have improved brightness, softness and core stability.