Order Entry
United States
ContactUsLinkComponent
Michel Applying and Removing Forceps, OR Grade, Sklar
Michel Applying and Removing Forceps, OR Grade, Sklar
Catalog # 10195-402
Supplier:  Sklar
undefined
Michel Applying and Removing Forceps, OR Grade, Sklar
Catalog # 10195-402
Supplier:  Sklar
Supplier Number:  24-1250

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Length:
    127 mm (5")
  • Material:
    Stainless steel
  • Cat. No.:
    10195-402
  • Jaw type:
    Straight
  • Tip style:
    Blunt, Serrated
  • Supplier no.:
    24-1250

Specifications

About this item

Michel Applying and Removing Forceps are thumb forceps, with a single spring mechanism, used to apply and remove surgical clips.

Clips may be applied to wounds and removed when healing is complete or they may be used as part of a procedure to stop blood flow.

This instrument has serrated edges, so the clip can be gripped for proper placement and secure removal.