HAZWIK® SOCs Socks, Brady®
Supplier: Brady Worldwide
Certificates
About this item
Dependable, flexible tube that contains and absorb liquids
Molds around corners and conforms to uneven surfaces. Available in a wide variety of types and sizes. The polypropylene SOC preferred when aggressive fluids are a problem. Color-coded green for safety, as well as wringable and incinerable.
Specifications
- Catalog No:
- 10796-182
- 10796-180
- Class Type:
- Chemical
- Chemical
- Color:
- Green
- Green
Specifications
Frequently Bought Together