Mr. Clean® Magic Eraser® Extra Power Cleaning Pad
102866-044
- Description:Scrub Sponge
- Thickness:0.7"
- Cat. No.:102866-044
- Supplier no.:PGC16449
Innovative water-activated micro-scrubbers reach into surface grooves, lifting away the toughest soils with water alone.
- Foam material
- Heavy duty rating
- Wavy rectangle shape
- White
Textured and 50% stronger than Magic Eraser® Original for heavy-duty cleaning. Holds up to tough messes better and lasts longer before you need to replace it. Ideal for use on kitchen appliances, tiles and grout, and even grill grates, lawn furniture and more.