H-D-Pro-Pro-Glu-NH2 Trifluoroacetate
Supplier: Bachem Americas
Certificates
About this item
In the presence of as little as 1 mol% pPE-amide, a broad range of aldehydes and nitroolefins react readily with each other. The resulting γ-nitroaldehydes were obtained in excellent yields and stereoselectivities at room temperature.
Specifications
- Source:
- Species:
- Environmentally Preferable:
Specifications
Frequently Bought Together