ContactUsLinkComponent
Peet Splinter Forceps, OR Grade, Sklar
  10195-030
 :  Sklar
undefined
Peet Splinter Forceps, OR Grade, Sklar
  10195-030
 :  Sklar
 :  19-3147
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Length:
    121 mm (4¾")
  • Cat. No.:
    10195-030
  • Jaw type:
    Serrated
  • Tip style:
    Double, 'S' Curved
  • Supplier no.:
    19-3147
Peet Splinter Forceps are used for placement or removal of objects
They are a ratcheted finger ring forceps, that provide a secure grip. Peet forceps feature a double curve and are used in Endodontics because the jaw allows a great view of the area inside the mouth.
They may also be known as Peet silver point forceps as they are used in endodontics to remove silver cones.