About this item
BDNF is a member of the NGF family of neurotrophic growth factors. Like other members of this family, BDNF supports neuron proliferation and survival. BDNF can bind to a low affinity cell surface receptor called LNGFR, which also binds other neurotrophins such as NGF, NT-3 and NT-4. However, BDNF mediates its neurotrophic properties by signaling through a high affinity cell surface receptor called gp145/trkB. BDNF is expressed as the C-terminal portion of a 247 amino acid polypeptide precursor, which also contains a signal sequence of 18 amino acid residues and a propeptide of 110 amino acid residues. Recombinant Human BDNF is a 27.0 kDa homodimer of two 120 amino acid subunits linked by strong non-covalent interactions. Human and Mouse BDNF sequences are identical. Manufactured using all Animal-Free reagents.
- Carrier free
- Animal-free
MHSDPARRGE LSVCDSISEW VTAADKKTAV DMSGGTVTVL EKVPVSKGQL KQYFYETKCN PMGYTKEGCR GIDKRHWNSQ CRTTQSYVRA LTMDSKKRIG WRFIRIDTSC VCTLTIKRGR
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:−20 °C
- Endotoxin Content:<0.1 ng/μg of protein (<1 EU/μg).
- Biological Activity:Determined by its ability to stimulate the proliferation of rat C6 cells. The expected ED50 for this effect is 0.5-1.0 μg/ml.
- Animal-Free:Y
- Protein/Peptide Name:BDNF
- Purity:Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Formulation:Lyophilized
- Shipping Temperature:RT