Mouse Recombinant TRAIL (from E. coli)
About this item
TRAIL is a cytotoxic protein, which activates rapid apoptosis in tumor cells, but not in normal cells. TRAIL-induced apoptosis is achieved through binding to two death-signaling receptors, DR4 and DR5. These receptors belong to the TNFR superfamily of transmembrane proteins, and contain a cytoplasmic “death domain”, which activates the cell’s apoptotic machinery. Recombinant Murine TRAIL is a 174 amino acid polypeptide (20.0 kDa), consisting of the TNF-homologous portion of the extracellular domain of the full length TRAIL protein.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
6 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
TRAIL is a cytotoxic protein, which activates rapid apoptosis in tumor cells, but not in normal cells. TRAIL-induced apoptosis is achieved through binding to two death-signaling receptors, DR4 and DR5. These receptors belong to the TNFR superfamily of transmembrane proteins, and contain a cytoplasmic “death domain”, which activates the cell’s apoptotic machinery. Recombinant Murine TRAIL is a 174 amino acid polypeptide (20.0 kDa), consisting of the TNF-homologous portion of the extracellular domain of the full length TRAIL protein.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 10780-782
- 10780-784
- 10780-786
- 10780-788
- 10780-790
- 10780-792
- Protein/Peptide Name:
- TRAIL
- TRAIL
- TRAIL
- TRAIL
- TRAIL
- TRAIL
- Protein Synonyms:
- TNF-Related Apoptosis-Inducing Ligand|CD253|TL2|TNFSF10|Apo2 Ligand
- TNF-Related Apoptosis-Inducing Ligand|CD253|TL2|TNFSF10|Apo2 Ligand
- TNF-Related Apoptosis-Inducing Ligand|CD253|TL2|TNFSF10|Apo2 Ligand
- TNF-Related Apoptosis-Inducing Ligand|CD253|TL2|TNFSF10|Apo2 Ligand
- TNF-Related Apoptosis-Inducing Ligand|CD253|TL2|TNFSF10|Apo2 Ligand
- TNF-Related Apoptosis-Inducing Ligand|CD253|TL2|TNFSF10|Apo2 Ligand
- Protein/Peptide Type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Species:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- 22035 P50592
- 22035 P50592
- 22035 P50592
- 22035 P50592
- 22035 P50592
- 22035 P50592
- Sequence:
- MRGGRPQKVA AHITGITRRS NSALIPISKD GKTLGQKIES WESSRKGHSF LNHVLFRNGE LVIEQEGLYY IYSQTYFRFQ EAEDASKMVS KDKVRTKQLV QYIYKYTSYP DPIVLMKSAR NSCWSRDAEY GLYSIYQGGL FELKKNDRIF VSVTNEHLMD LDQEASFFGA FLIN
- MRGGRPQKVA AHITGITRRS NSALIPISKD GKTLGQKIES WESSRKGHSF LNHVLFRNGE LVIEQEGLYY IYSQTYFRFQ EAEDASKMVS KDKVRTKQLV QYIYKYTSYP DPIVLMKSAR NSCWSRDAEY GLYSIYQGGL FELKKNDRIF VSVTNEHLMD LDQEASFFGA FLIN
- MRGGRPQKVA AHITGITRRS NSALIPISKD GKTLGQKIES WESSRKGHSF LNHVLFRNGE LVIEQEGLYY IYSQTYFRFQ EAEDASKMVS KDKVRTKQLV QYIYKYTSYP DPIVLMKSAR NSCWSRDAEY GLYSIYQGGL FELKKNDRIF VSVTNEHLMD LDQEASFFGA FLIN
- MRGGRPQKVA AHITGITRRS NSALIPISKD GKTLGQKIES WESSRKGHSF LNHVLFRNGE LVIEQEGLYY IYSQTYFRFQ EAEDASKMVS KDKVRTKQLV QYIYKYTSYP DPIVLMKSAR NSCWSRDAEY GLYSIYQGGL FELKKNDRIF VSVTNEHLMD LDQEASFFGA FLIN
- MRGGRPQKVA AHITGITRRS NSALIPISKD GKTLGQKIES WESSRKGHSF LNHVLFRNGE LVIEQEGLYY IYSQTYFRFQ EAEDASKMVS KDKVRTKQLV QYIYKYTSYP DPIVLMKSAR NSCWSRDAEY GLYSIYQGGL FELKKNDRIF VSVTNEHLMD LDQEASFFGA FLIN
- MRGGRPQKVA AHITGITRRS NSALIPISKD GKTLGQKIES WESSRKGHSF LNHVLFRNGE LVIEQEGLYY IYSQTYFRFQ EAEDASKMVS KDKVRTKQLV QYIYKYTSYP DPIVLMKSAR NSCWSRDAEY GLYSIYQGGL FELKKNDRIF VSVTNEHLMD LDQEASFFGA FLIN
- Protease-free:
- Y
- Y
- Y
- Y
- Y
- Y
- Endotoxin Level:
- Low
- Low
- Low
- Low
- Low
- Low
- Endotoxin Content:
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- Biological Activity:
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Purity:
- 0.95
- 0.95
- 0.95
- 0.95
- 0.95
- 0.95
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Formulation:
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Molecular Weight:
- 20 kDa
- 20 kDa
- 20 kDa
- 20 kDa
- 20 kDa
- 20 kDa
- Reconstitution Instructions:
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Storage Conditions:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping Temperature:
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
Specifications
Product Family Options
Product Information
- SizeOriginAvailabilityPrice
- Supplier Number: 315-19-10UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10780-782Protein/Peptide NameTRAILProtein SynonymsTNF-Related Apoptosis-Inducing Ligand|CD253|TL2|TNFSF10|Apo2 LigandProtein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID22035 P50592SequenceMRGGRPQKVA AHITGITRRS NSALIPISKD GKTLGQKIES WESSRKGHSF LNHVLFRNGE LVIEQEGLYY IYSQTYFRFQ EAEDASKMVS KDKVRTKQLV QYIYKYTSYP DPIVLMKSAR NSCWSRDAEY GLYSIYQGGL FELKKNDRIF VSVTNEHLMD LDQEASFFGA FLINProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.95FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight20 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: 315-19-50UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10780-784Protein/Peptide NameTRAILProtein SynonymsTNF-Related Apoptosis-Inducing Ligand|CD253|TL2|TNFSF10|Apo2 LigandProtein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID22035 P50592SequenceMRGGRPQKVA AHITGITRRS NSALIPISKD GKTLGQKIES WESSRKGHSF LNHVLFRNGE LVIEQEGLYY IYSQTYFRFQ EAEDASKMVS KDKVRTKQLV QYIYKYTSYP DPIVLMKSAR NSCWSRDAEY GLYSIYQGGL FELKKNDRIF VSVTNEHLMD LDQEASFFGA FLINProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.95FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight20 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: 315-19-100UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10780-786Protein/Peptide NameTRAILProtein SynonymsTNF-Related Apoptosis-Inducing Ligand|CD253|TL2|TNFSF10|Apo2 LigandProtein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID22035 P50592SequenceMRGGRPQKVA AHITGITRRS NSALIPISKD GKTLGQKIES WESSRKGHSF LNHVLFRNGE LVIEQEGLYY IYSQTYFRFQ EAEDASKMVS KDKVRTKQLV QYIYKYTSYP DPIVLMKSAR NSCWSRDAEY GLYSIYQGGL FELKKNDRIF VSVTNEHLMD LDQEASFFGA FLINProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.95FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight20 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: 315-19-250UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10780-788Protein/Peptide NameTRAILProtein SynonymsTNF-Related Apoptosis-Inducing Ligand|CD253|TL2|TNFSF10|Apo2 LigandProtein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID22035 P50592SequenceMRGGRPQKVA AHITGITRRS NSALIPISKD GKTLGQKIES WESSRKGHSF LNHVLFRNGE LVIEQEGLYY IYSQTYFRFQ EAEDASKMVS KDKVRTKQLV QYIYKYTSYP DPIVLMKSAR NSCWSRDAEY GLYSIYQGGL FELKKNDRIF VSVTNEHLMD LDQEASFFGA FLINProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.95FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight20 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: 315-19-500UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10780-790Protein/Peptide NameTRAILProtein SynonymsTNF-Related Apoptosis-Inducing Ligand|CD253|TL2|TNFSF10|Apo2 LigandProtein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID22035 P50592SequenceMRGGRPQKVA AHITGITRRS NSALIPISKD GKTLGQKIES WESSRKGHSF LNHVLFRNGE LVIEQEGLYY IYSQTYFRFQ EAEDASKMVS KDKVRTKQLV QYIYKYTSYP DPIVLMKSAR NSCWSRDAEY GLYSIYQGGL FELKKNDRIF VSVTNEHLMD LDQEASFFGA FLINProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.95FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight20 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: 315-19-1MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10780-792Protein/Peptide NameTRAILProtein SynonymsTNF-Related Apoptosis-Inducing Ligand|CD253|TL2|TNFSF10|Apo2 LigandProtein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID22035 P50592SequenceMRGGRPQKVA AHITGITRRS NSALIPISKD GKTLGQKIES WESSRKGHSF LNHVLFRNGE LVIEQEGLYY IYSQTYFRFQ EAEDASKMVS KDKVRTKQLV QYIYKYTSYP DPIVLMKSAR NSCWSRDAEY GLYSIYQGGL FELKKNDRIF VSVTNEHLMD LDQEASFFGA FLINProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.95FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight20 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



