Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant TWEAK (from E. coli)
 :  Peprotech
 
Human Recombinant TWEAK (from E. coli)
 :  Peprotech

 

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <0.1 ng/μg of protein (<1 EU/μg).
  • Biological Activity:
    Assay #1: The ED50 as determined by the dose-dependent stimulation of IL-8 production by human PBMC is less than 10 ng/ml.
    Assay #2: TWEAK weakly induces the death of HT29 cells when cultured in the presence of IFN-γ. The ED50 for this effect is between 30-45 ng/ml.
  • Gene ID:
    8742
  • Animal-Free:
    Y
  • Protein/Peptide Name:
    TWEAK
  • Purity:
    Greater than 98% by SDS-PAGE gel and HPLC analyses.
  • Formulation:
    Lyophilized
  • Shipping Temperature:
    RT

 

Frequently Bought Together