Human Recombinant BD-5 (from E. coli)
About this item
Defensins (α and β) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The α-defensins are distinguished from the β-defensins by the pairing of their three disulfide bonds. To date, six human β-defensins have been identified; BD-1, BD-2, BD-3, BD-4, BD-5 and BD-6. β-defensins are expressed on some leukocytes and at epithelial surfaces. In addition to their direct antimicrobial activities, they can act as chemoattractants towards immature dendritic cells and memory T cells. The β-defensin proteins are expressed as the C-terminal portion of precursors, and are released by proteolytic cleavage of a signal sequence and, in some cases, a propeptide sequence. β-defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds. Recombinant Human BD-5 is a 5.8 kDa protein containing 51 amino acid residues.
- Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
6 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Defensins (α and β) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The α-defensins are distinguished from the β-defensins by the pairing of their three disulfide bonds. To date, six human β-defensins have been identified; BD-1, BD-2, BD-3, BD-4, BD-5 and BD-6. β-defensins are expressed on some leukocytes and at epithelial surfaces. In addition to their direct antimicrobial activities, they can act as chemoattractants towards immature dendritic cells and memory T cells. The β-defensin proteins are expressed as the C-terminal portion of precursors, and are released by proteolytic cleavage of a signal sequence and, in some cases, a propeptide sequence. β-defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds. Recombinant Human BD-5 is a 5.8 kDa protein containing 51 amino acid residues.
- Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 10778-724
- 10778-726
- 10778-728
- 10778-730
- 10778-732
- 10778-734
- Protein/Peptide Name:
- BD-5
- BD-5
- BD-5
- BD-5
- BD-5
- BD-5
- Protein Synonyms:
- DEFB5|Beta-Defensin 5|BD5|DEFB105a
- DEFB5|Beta-Defensin 5|BD5|DEFB105a
- DEFB5|Beta-Defensin 5|BD5|DEFB105a
- DEFB5|Beta-Defensin 5|BD5|DEFB105a
- DEFB5|Beta-Defensin 5|BD5|DEFB105a
- DEFB5|Beta-Defensin 5|BD5|DEFB105a
- Protein/Peptide Type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Species:
- Human
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- 245908 Q8NG35
- 245908 Q8NG35
- 245908 Q8NG35
- 245908 Q8NG35
- 245908 Q8NG35
- 245908 Q8NG35
- Sequence:
- GLDFSQPFPS GEFAVCESCK LGRGKCRKEC LENEKPDGNC RLNFLCCRQR I
- GLDFSQPFPS GEFAVCESCK LGRGKCRKEC LENEKPDGNC RLNFLCCRQR I
- GLDFSQPFPS GEFAVCESCK LGRGKCRKEC LENEKPDGNC RLNFLCCRQR I
- GLDFSQPFPS GEFAVCESCK LGRGKCRKEC LENEKPDGNC RLNFLCCRQR I
- GLDFSQPFPS GEFAVCESCK LGRGKCRKEC LENEKPDGNC RLNFLCCRQR I
- GLDFSQPFPS GEFAVCESCK LGRGKCRKEC LENEKPDGNC RLNFLCCRQR I
- Protease-free:
- Y
- Y
- Y
- Y
- Y
- Y
- Animal-Free:
- Y
- Y
- Y
- Y
- Y
- Y
- Endotoxin Level:
- Low
- Low
- Low
- Low
- Low
- Low
- Endotoxin Content:
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- Biological Activity:
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Purity:
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Formulation:
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Molecular Weight:
- 5.8 kDa
- 5.8 kDa
- 5.8 kDa
- 5.8 kDa
- 5.8 kDa
- 5.8 kDa
- Reconstitution Instructions:
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Storage Conditions:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping Temperature:
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
Specifications
Product Family Options
Product Information
- SizeOriginAvailabilityPrice
- Supplier Number: AF-300-68-5UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-724Protein/Peptide NameBD-5Protein SynonymsDEFB5|Beta-Defensin 5|BD5|DEFB105aProtein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID245908 Q8NG35SequenceGLDFSQPFPS GEFAVCESCK LGRGKCRKEC LENEKPDGNC RLNFLCCRQR IProtease-freeYAnimal-FreeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight5.8 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: AF-300-68-20UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-726Protein/Peptide NameBD-5Protein SynonymsDEFB5|Beta-Defensin 5|BD5|DEFB105aProtein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID245908 Q8NG35SequenceGLDFSQPFPS GEFAVCESCK LGRGKCRKEC LENEKPDGNC RLNFLCCRQR IProtease-freeYAnimal-FreeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight5.8 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: AF-300-68-100UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-728Protein/Peptide NameBD-5Protein SynonymsDEFB5|Beta-Defensin 5|BD5|DEFB105aProtein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID245908 Q8NG35SequenceGLDFSQPFPS GEFAVCESCK LGRGKCRKEC LENEKPDGNC RLNFLCCRQR IProtease-freeYAnimal-FreeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight5.8 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: AF-300-68-250UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-730Protein/Peptide NameBD-5Protein SynonymsDEFB5|Beta-Defensin 5|BD5|DEFB105aProtein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID245908 Q8NG35SequenceGLDFSQPFPS GEFAVCESCK LGRGKCRKEC LENEKPDGNC RLNFLCCRQR IProtease-freeYAnimal-FreeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight5.8 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: AF-300-68-500UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-732Protein/Peptide NameBD-5Protein SynonymsDEFB5|Beta-Defensin 5|BD5|DEFB105aProtein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID245908 Q8NG35SequenceGLDFSQPFPS GEFAVCESCK LGRGKCRKEC LENEKPDGNC RLNFLCCRQR IProtease-freeYAnimal-FreeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight5.8 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: AF-300-68-1MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10778-734Protein/Peptide NameBD-5Protein SynonymsDEFB5|Beta-Defensin 5|BD5|DEFB105aProtein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID245908 Q8NG35SequenceGLDFSQPFPS GEFAVCESCK LGRGKCRKEC LENEKPDGNC RLNFLCCRQR IProtease-freeYAnimal-FreeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight5.8 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



