Order Entry
United States
Orders LinkContactUsLinkComponent
 

Mouse Recombinant IL-4 (from E. coli)

Supplier:   Peprotech
Mouse Recombinant IL-4 (from <i>E. coli</i>)
Supplier:  Peprotech
Certificates

7 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

IL-4 is a pleiotropic cytokine that regulates diverse T and B cell responses including cell proliferation, survival and gene expression. Produced by mast cells, T cells and bone marrow stromal cells, IL-4 regulates the differentiation of naive CD4+ T cells into helper Th2 cells, characterized by their cytokine-secretion profile that includes secretion of IL-4, IL-5, IL-6, IL-10, and IL-13, which favor a humoral immune response. Another dominant function of IL-4 is the regulation of immunoglobulin class switching to the IgG1 and IgE isotypes. Excessive IL-4 production by Th2 cells has been associated with elevated IgE production and allergy. Recombinant Murine IL-4 is a 13.5 kDa globular protein containing 121 amino acid residues.

  • Animal-free

 : For research use only. Not for use in diagnostic or therapeutic procedures.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 10778-476
  • 10778-478
  • 10778-480
  • 10778-482
  • 10778-484
  • 10778-486
  • 10778-488
  • Protein/Peptide Name:
  • IL-4
  • IL-4
  • IL-4
  • IL-4
  • IL-4
  • IL-4
  • IL-4
  • Protein Synonyms:
  • B-cell Stimulating Factor (BSF-1)|BCGF|BCDF|Interleukin-4
  • B-cell Stimulating Factor (BSF-1)|BCGF|BCDF|Interleukin-4
  • B-cell Stimulating Factor (BSF-1)|BCGF|BCDF|Interleukin-4
  • B-cell Stimulating Factor (BSF-1)|BCGF|BCDF|Interleukin-4
  • B-cell Stimulating Factor (BSF-1)|BCGF|BCDF|Interleukin-4
  • B-cell Stimulating Factor (BSF-1)|BCGF|BCDF|Interleukin-4
  • B-cell Stimulating Factor (BSF-1)|BCGF|BCDF|Interleukin-4
  • Protein/Peptide Type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein Function:
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Species:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • 16189 P07750
  • 16189 P07750
  • 16189 P07750
  • 16189 P07750
  • 16189 P07750
  • 16189 P07750
  • 16189 P07750
  • Sequence:
  • MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S
  • MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S
  • MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S
  • MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S
  • MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S
  • MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S
  • MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S
  • Protease-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Y
  • Y
  • Animal-Free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin Level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin Content:
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • Biological Activity:
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Purity:
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • Form:
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Formulation:
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Molecular Weight:
  • 13.5 kDa
  • 13.5 kDa
  • 13.5 kDa
  • 13.5 kDa
  • 13.5 kDa
  • 13.5 kDa
  • 13.5 kDa
  • Reconstitution Instructions:
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Storage Conditions:
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Shipping Temperature:
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient

Specifications

Product Family Options

Product Information

  • Size
    Origin
    Availability
    Price
  • 5 μg
    US
    Log in for availability
    Log in for availability
    Each (5µG)
    Each (5µG) - $199.54
    $199.54
    Each (5µG)
       $199.54
     

    Supplier Number: AF-214-14-5UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10778-476
    Protein/Peptide Name
    IL-4
    Protein Synonyms
    B-cell Stimulating Factor (BSF-1)|BCGF|BCDF|Interleukin-4
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    16189 P07750
    Sequence
    MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S
    Protease-free
    Y
    Animal-Free
    Y
    Endotoxin Level
    Low
    Endotoxin Content
    >0.1 ng/μg
    Biological Activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular Weight
    13.5 kDa
    Reconstitution Instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage Conditions
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient
  • 20 μg
    US
    Log in for availability
    Log in for availability
    Each (20µG)
    Each (20µG) - $446.75
    $446.75
    Each (20µG)
       $446.75
     

    Supplier Number: AF-214-14-20UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10778-478
    Protein/Peptide Name
    IL-4
    Protein Synonyms
    B-cell Stimulating Factor (BSF-1)|BCGF|BCDF|Interleukin-4
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    16189 P07750
    Sequence
    MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S
    Protease-free
    Y
    Animal-Free
    Y
    Endotoxin Level
    Low
    Endotoxin Content
    >0.1 ng/μg
    Biological Activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular Weight
    13.5 kDa
    Reconstitution Instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage Conditions
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient
  • 50 μg
    US
    Log in for availability
    Log in for availability
    Each (50µG)
    Each (50µG) - $857.19
    $857.19
    Each (50µG)
       $857.19
     

    Supplier Number: AF-214-14-50UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10778-480
    Protein/Peptide Name
    IL-4
    Protein Synonyms
    B-cell Stimulating Factor (BSF-1)|BCGF|BCDF|Interleukin-4
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    16189 P07750
    Sequence
    MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S
    Protease-free
    Y
    Animal-Free
    Y
    Endotoxin Level
    Low
    Endotoxin Content
    >0.1 ng/μg
    Biological Activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular Weight
    13.5 kDa
    Reconstitution Instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage Conditions
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient
  • 100 μg
    US
    Log in for availability
    Log in for availability
    Each (100µG)
    Each (100µG) - $1,341.07
    $1,341.07
    Each (100µG)
       $1,341.07
     

    Supplier Number: AF-214-14-100UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10778-482
    Protein/Peptide Name
    IL-4
    Protein Synonyms
    B-cell Stimulating Factor (BSF-1)|BCGF|BCDF|Interleukin-4
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    16189 P07750
    Sequence
    MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S
    Protease-free
    Y
    Animal-Free
    Y
    Endotoxin Level
    Low
    Endotoxin Content
    >0.1 ng/μg
    Biological Activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular Weight
    13.5 kDa
    Reconstitution Instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage Conditions
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient
  • 250 μg
    US
    Log in for availability
    Log in for availability
    Each (250µG)
    Each (250µG) - $2,142.81
    $2,142.81
    Each (250µG)
       $2,142.81
     

    Supplier Number: AF-214-14-250UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10778-484
    Protein/Peptide Name
    IL-4
    Protein Synonyms
    B-cell Stimulating Factor (BSF-1)|BCGF|BCDF|Interleukin-4
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    16189 P07750
    Sequence
    MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S
    Protease-free
    Y
    Animal-Free
    Y
    Endotoxin Level
    Low
    Endotoxin Content
    >0.1 ng/μg
    Biological Activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular Weight
    13.5 kDa
    Reconstitution Instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage Conditions
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient
  • 500 μg
    US
    Log in for availability
    Log in for availability
    Each (500µG)
    Each (500µG) - $3,518.15
    $3,518.15
    Each (500µG)
       $3,518.15
     

    Supplier Number: AF-214-14-500UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10778-486
    Protein/Peptide Name
    IL-4
    Protein Synonyms
    B-cell Stimulating Factor (BSF-1)|BCGF|BCDF|Interleukin-4
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    16189 P07750
    Sequence
    MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S
    Protease-free
    Y
    Animal-Free
    Y
    Endotoxin Level
    Low
    Endotoxin Content
    >0.1 ng/μg
    Biological Activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular Weight
    13.5 kDa
    Reconstitution Instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage Conditions
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient
  • 1 mg
    US
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $5,865.91
    $5,865.91
    Each (1mg)
       $5,865.91
     

    Supplier Number: AF-214-14-1MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10778-488
    Protein/Peptide Name
    IL-4
    Protein Synonyms
    B-cell Stimulating Factor (BSF-1)|BCGF|BCDF|Interleukin-4
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    16189 P07750
    Sequence
    MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S
    Protease-free
    Y
    Animal-Free
    Y
    Endotoxin Level
    Low
    Endotoxin Content
    >0.1 ng/μg
    Biological Activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular Weight
    13.5 kDa
    Reconstitution Instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage Conditions
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient