Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant IL4 (from E. coli)
 :  Peprotech
 
Human Recombinant IL4 (from E. coli)
 :  Peprotech
 

 

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <0.1 ng/μg of protein (<1 EU/μg).
  • Biological Activity:
    The ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 0.2 ng/ml, corresponding to a specific activity of ≥ 5 x 106 units/mg.
  • Animal-Free:
    Y
  • Protein/Peptide Name:
    IL4
  • Purity:
    Greater than 98% by SDS-PAGE gel and HPLC analyses.
  • Formulation:
    Lyophilized
  • Shipping Temperature:
    RT

 

 

Frequently Bought Together