Order Entry
United States
ContactUsLinkComponent
Human Recombinant ApoA-I (from E. coli)
Human Recombinant ApoA-I (from E. coli)
Catalog # 10774-488
Supplier:  Peprotech
CAS Number:  
Human Recombinant ApoA-I (from E. coli)
Catalog # 10774-488
Supplier:  Peprotech
Supplier Number:  350-11-20UG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    20 μg
  • Storage Conditions:
    Store at –20 °C prior to reconstitution.
  • Endotoxin Content:
    >0.1 ng/μg
  • Biological Activity:
    Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Form:
    Lyophilized
  • Gene ID:
    335 P02647
  • Reconstitution Instructions:
    Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Protease-free:
    Y
  • Protein Synonyms:
    Apolipoprotein A-I
  • Protein/Peptide Name:
    ApoA-I
  • Purity:
    0.97
  • Molecular Weight:
    28.2 kDa
  • Sequence:
    MDEPPQSPWD RVKDLATVYV DVLKDSGRDY VSQFEGSALG KQLNLKLLDN WDSVTSTFSK LREQLGPVTQ EFWDNLEKET EGLRQEMSKD LEEVKAKVQP YLDDFQKKWQ EEMELYRQKV EPLRAELQEG ARQKLHELQE KLSPLGEEMR DRARAHVDAL RTHLAPYSDE LRQRLAARLE ALKENGGARL AEYHAKATEH LSTLSEKAKP ALEDLRQGLL PVLESFKVSF LSALEEYTKK LNTQ
  • Endotoxin Level:
    Low
  • Formulation:
    Formulation can vary. Please refer to product CoA for information.
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    10774-488
  • Origin:
    US
  • Supplier no.:
    350-11-20UG

Specifications

About this item

ApoA-I is a 29.0 kDa protein produced in the liver and intestine, and secreted as the predominant constituent of nascent high density lipoprotein (HDL) particle. ApoA-I, which is found exclusively in HDL, has a unique ability to capture and solubilize free cholesterol. This ApoA-I ability enables HDL to remove excess peripheral cholesterol, and return it to the liver for recycling and excretion. This process, called reverse cholesterol transport, is thought to inhibit atherogenesis. For this reason, HDL is also known as the “good cholesterol.” The therapeutic potential of ApoA-I has been recently assessed in patients with acute coronary syndromes, using a recombinant form of a naturally occurring variant of ApoA-I (called ApoA-I Milano). The availability of recombinant normal ApoA-I should facilitate further investigation into the potential usefulness of ApoA-I in preventing atherosclerotic vascular diseases. Recombinant Human ApoA-I is a 28.2 kDa protein of 244 amino acid residues.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.