Order Entry
United States
Orders LinkContactUsLinkComponent
Human Recombinant TWEAK (from <i>E. coli</i>)

Human Recombinant TWEAK (from E. coli)

Supplier:   Peprotech
Human Recombinant TWEAK (from <i>E. coli</i>)
Supplier:  Peprotech
Certificates

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

TWEAKR belongs to the TNF family of transmembrane proteins, and contains a cytoplasmic "death domain," which can activate a cell's apoptotic machinery. It is expressed in the spleen, thymus, peripheral blood lymphocytes, colon, and small intestine. Signal transduction by TWEAKR can be activated by either membrane-anchored or soluble TWEAK. Recombinant Human TWEAK Receptor is a 53 amino acid polypeptide (5.6 kDa) comprising the entire extracellular domain of the full-length TWEAKR protein.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 10774-318
  • 10774-320
  • Protein/Peptide Name:
  • TWEAK
  • TWEAK
  • Protein Synonyms:
  • FGF-inducible 14|FN14|TNF-related Weak Inducer of Apoptosis Receptor|TNFRSF12A
  • FGF-inducible 14|FN14|TNF-related Weak Inducer of Apoptosis Receptor|TNFRSF12A
  • Protein/Peptide Type:
  • Recombinant
  • Recombinant
  • Protein Function:
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Species:
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • 51330 Q9NP84
  • 51330 Q9NP84
  • Sequence:
  • EQAPGTAPCS RGSSWSADLD KCMDCASCRA RPHSDFCLGC AAAPPAPFRL LWP
  • EQAPGTAPCS RGSSWSADLD KCMDCASCRA RPHSDFCLGC AAAPPAPFRL LWP
  • Protease-free:
  • Y
  • Y
  • Endotoxin Level:
  • Low
  • Low
  • Endotoxin Content:
  • >0.1 ng/μg
  • >0.1 ng/μg
  • Biological Activity:
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Purity:
  • 0.98
  • 0.98
  • Form:
  • Lyophilized
  • Lyophilized
  • Formulation:
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Molecular Weight:
  • 5.6 kDa
  • 5.6 kDa
  • Reconstitution Instructions:
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Storage Conditions:
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Shipping Temperature:
  • Ambient
  • Ambient

Specifications

Product Family Options

Product Information

  • Size
    Origin
    Availability
    Price
  • 5 μg
    US
    Log in for availability
    Log in for availability
    Each (5µG)
    Each (5µG) - $144.91
    $144.91
    Each (5µG)
       $144.91
    Minimum order quantity is 1

    Supplier Number: 310-21-5UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10774-318
    Protein/Peptide Name
    TWEAK
    Protein Synonyms
    FGF-inducible 14|FN14|TNF-related Weak Inducer of Apoptosis Receptor|TNFRSF12A
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    51330 Q9NP84
    Sequence
    EQAPGTAPCS RGSSWSADLD KCMDCASCRA RPHSDFCLGC AAAPPAPFRL LWP
    Protease-free
    Y
    Endotoxin Level
    Low
    Endotoxin Content
    >0.1 ng/μg
    Biological Activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular Weight
    5.6 kDa
    Reconstitution Instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage Conditions
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient
  • 25 μg
    US
    Log in for availability
    Log in for availability
    Each (25µG)
    Each (25µG) - $353.43
    $353.43
    Each (25µG)
       $353.43
    Minimum order quantity is 1

    Supplier Number: 310-21-25UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10774-320
    Protein/Peptide Name
    TWEAK
    Protein Synonyms
    FGF-inducible 14|FN14|TNF-related Weak Inducer of Apoptosis Receptor|TNFRSF12A
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    51330 Q9NP84
    Sequence
    EQAPGTAPCS RGSSWSADLD KCMDCASCRA RPHSDFCLGC AAAPPAPFRL LWP
    Protease-free
    Y
    Endotoxin Level
    Low
    Endotoxin Content
    >0.1 ng/μg
    Biological Activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular Weight
    5.6 kDa
    Reconstitution Instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage Conditions
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.