Human Recombinant Eotaxin-2 (from E. coli)
About this item
Eotaxin-2 is a CC chemokine that signals through the CCR3 receptor. It is produced by activated monocytes and T lymphocytes. Eotaxin-2 selectively chemoattracts cells expressing CCR3, including eosinophils, basophils, Th2 T cells, mast cells, and certain subsets of dendritic cells. Additionally, Eotaxin-2 inhibits the proliferation of multipotential hematopoietic progenitor cells. The mature protein, which also includes a C-terminal truncation, contains 78 amino acid residues (92 a.a. residues for the murine homolog, without C-terminal truncation). Eotaxin-2 contains the four conserved cysteine residues present in CC chemokines. Recombinant Human Eotaxin-2 (CCL24) is an 8.8 kDa protein containing 78 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
7 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Eotaxin-2 is a CC chemokine that signals through the CCR3 receptor. It is produced by activated monocytes and T lymphocytes. Eotaxin-2 selectively chemoattracts cells expressing CCR3, including eosinophils, basophils, Th2 T cells, mast cells, and certain subsets of dendritic cells. Additionally, Eotaxin-2 inhibits the proliferation of multipotential hematopoietic progenitor cells. The mature protein, which also includes a C-terminal truncation, contains 78 amino acid residues (92 a.a. residues for the murine homolog, without C-terminal truncation). Eotaxin-2 contains the four conserved cysteine residues present in CC chemokines. Recombinant Human Eotaxin-2 (CCL24) is an 8.8 kDa protein containing 78 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 10773-830
- 10773-832
- 10773-834
- 10773-836
- 10773-838
- 10773-840
- 10773-842
- Protein/Peptide Name:
- Eotaxin-2
- Eotaxin-2
- Eotaxin-2
- Eotaxin-2
- Eotaxin-2
- Eotaxin-2
- Eotaxin-2
- Protein Synonyms:
- MPIF-2|CkB-6
- MPIF-2|CkB-6
- MPIF-2|CkB-6
- MPIF-2|CkB-6
- MPIF-2|CkB-6
- MPIF-2|CkB-6
- MPIF-2|CkB-6
- Protein/Peptide Type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Species:
- Human
- Human
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- 6369 O00175
- 6369 O00175
- 6369 O00175
- 6369 O00175
- 6369 O00175
- 6369 O00175
- 6369 O00175
- Sequence:
- VVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVA
- VVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVA
- VVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVA
- VVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVA
- VVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVA
- VVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVA
- VVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVA
- Protease-free:
- Y
- Y
- Y
- Y
- Y
- Y
- Y
- Endotoxin Level:
- Low
- Low
- Low
- Low
- Low
- Low
- Low
- Endotoxin Content:
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- Biological Activity:
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Purity:
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Formulation:
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Molecular Weight:
- 8.8 kDa
- 8.8 kDa
- 8.8 kDa
- 8.8 kDa
- 8.8 kDa
- 8.8 kDa
- 8.8 kDa
- Reconstitution Instructions:
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Storage Conditions:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping Temperature:
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
Specifications
Product Family Options
Product Information
- SizeOriginAvailabilityPrice
- Supplier Number: 300-33-5UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10773-830Protein/Peptide NameEotaxin-2Protein SynonymsMPIF-2|CkB-6Protein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID6369 O00175SequenceVVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVAProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight8.8 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: 300-33-20UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10773-832Protein/Peptide NameEotaxin-2Protein SynonymsMPIF-2|CkB-6Protein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID6369 O00175SequenceVVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVAProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight8.8 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: 300-33-50UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10773-834Protein/Peptide NameEotaxin-2Protein SynonymsMPIF-2|CkB-6Protein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID6369 O00175SequenceVVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVAProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight8.8 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: 300-33-100UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10773-836Protein/Peptide NameEotaxin-2Protein SynonymsMPIF-2|CkB-6Protein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID6369 O00175SequenceVVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVAProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight8.8 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: 300-33-250UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10773-838Protein/Peptide NameEotaxin-2Protein SynonymsMPIF-2|CkB-6Protein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID6369 O00175SequenceVVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVAProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight8.8 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: 300-33-500UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10773-840Protein/Peptide NameEotaxin-2Protein SynonymsMPIF-2|CkB-6Protein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID6369 O00175SequenceVVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVAProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight8.8 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: 300-33-1MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10773-842Protein/Peptide NameEotaxin-2Protein SynonymsMPIF-2|CkB-6Protein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID6369 O00175SequenceVVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVAProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight8.8 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



