Order Entry
United States
Orders LinkContactUsLinkComponent
 

Mouse Recombinant MDC (from E. coli)

Supplier:   Peprotech
Mouse Recombinant MDC (from <i>E. coli</i>)
Supplier:  Peprotech
Certificates

6 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

MDC is a CC chemokine that is produced in B cells, macrophages, monocyte-derived dendritic cells, activated NK cells and CD4 T cells. It signals through the CCR4 receptor. MDC chemoattracts monocytes, dendritic cells and NK cells, and exerts HIV-suppressive activity. The 67 amino acid form of MDC displays reduced chemoattractant activity, but retains HIV-suppressive activity. Recombinant Murine MDC (CCL22) is a 7.8 kDa protein containing 68 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines.

 : For research use only. Not for use in diagnostic or therapeutic procedures.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 10773-304
  • 10773-306
  • 10773-308
  • 10773-310
  • 10773-312
  • 10773-314
  • Protein/Peptide Name:
  • MDC
  • MDC
  • MDC
  • MDC
  • MDC
  • MDC
  • Protein Synonyms:
  • ABCD-1|Macrophage-Derived Chemokine|STCP-1
  • ABCD-1|Macrophage-Derived Chemokine|STCP-1
  • ABCD-1|Macrophage-Derived Chemokine|STCP-1
  • ABCD-1|Macrophage-Derived Chemokine|STCP-1
  • ABCD-1|Macrophage-Derived Chemokine|STCP-1
  • ABCD-1|Macrophage-Derived Chemokine|STCP-1
  • Protein/Peptide Type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein Function:
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Species:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • 20299 O88430
  • 20299 O88430
  • 20299 O88430
  • 20299 O88430
  • 20299 O88430
  • 20299 O88430
  • Sequence:
  • GPYGANVEDS ICCQDYIRHP LPSRLVKEFF WTSKSCRKPG VVLITVKNRD ICADPRQVWV KKLLHKLS
  • GPYGANVEDS ICCQDYIRHP LPSRLVKEFF WTSKSCRKPG VVLITVKNRD ICADPRQVWV KKLLHKLS
  • GPYGANVEDS ICCQDYIRHP LPSRLVKEFF WTSKSCRKPG VVLITVKNRD ICADPRQVWV KKLLHKLS
  • GPYGANVEDS ICCQDYIRHP LPSRLVKEFF WTSKSCRKPG VVLITVKNRD ICADPRQVWV KKLLHKLS
  • GPYGANVEDS ICCQDYIRHP LPSRLVKEFF WTSKSCRKPG VVLITVKNRD ICADPRQVWV KKLLHKLS
  • GPYGANVEDS ICCQDYIRHP LPSRLVKEFF WTSKSCRKPG VVLITVKNRD ICADPRQVWV KKLLHKLS
  • Protease-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin Level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin Content:
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • Biological Activity:
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Purity:
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • Form:
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Formulation:
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Molecular Weight:
  • 7.8 kDa
  • 7.8 kDa
  • 7.8 kDa
  • 7.8 kDa
  • 7.8 kDa
  • 7.8 kDa
  • Reconstitution Instructions:
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Storage Conditions:
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Shipping Temperature:
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient

Specifications

Product Family Options

Product Information

  • Size
    Origin
    Availability
    Price
  • 5 μg
    US
    Log in for availability
    Log in for availability
    Each (5µG)
    Each (5µG) - $144.91
    $144.91
    Each (5µG)
       $144.91
     

    Supplier Number: 250-23-5UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10773-304
    Protein/Peptide Name
    MDC
    Protein Synonyms
    ABCD-1|Macrophage-Derived Chemokine|STCP-1
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    20299 O88430
    Sequence
    GPYGANVEDS ICCQDYIRHP LPSRLVKEFF WTSKSCRKPG VVLITVKNRD ICADPRQVWV KKLLHKLS
    Protease-free
    Y
    Endotoxin Level
    Low
    Endotoxin Content
    >0.1 ng/μg
    Biological Activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular Weight
    7.8 kDa
    Reconstitution Instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage Conditions
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient
  • 20 μg
    US
    Log in for availability
    Log in for availability
    Each (20µG)
    Each (20µG) - $372.33
    $372.33
    Each (20µG)
       $372.33
     

    Supplier Number: 250-23-20UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10773-306
    Protein/Peptide Name
    MDC
    Protein Synonyms
    ABCD-1|Macrophage-Derived Chemokine|STCP-1
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    20299 O88430
    Sequence
    GPYGANVEDS ICCQDYIRHP LPSRLVKEFF WTSKSCRKPG VVLITVKNRD ICADPRQVWV KKLLHKLS
    Protease-free
    Y
    Endotoxin Level
    Low
    Endotoxin Content
    >0.1 ng/μg
    Biological Activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular Weight
    7.8 kDa
    Reconstitution Instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage Conditions
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient
  • 100 μg
    US
    Log in for availability
    Log in for availability
    Each (100µG)
    Each (100µG) - $1,083.40
    $1,083.40
    Each (100µG)
       $1,083.40
     

    Supplier Number: 250-23-100UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10773-308
    Protein/Peptide Name
    MDC
    Protein Synonyms
    ABCD-1|Macrophage-Derived Chemokine|STCP-1
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    20299 O88430
    Sequence
    GPYGANVEDS ICCQDYIRHP LPSRLVKEFF WTSKSCRKPG VVLITVKNRD ICADPRQVWV KKLLHKLS
    Protease-free
    Y
    Endotoxin Level
    Low
    Endotoxin Content
    >0.1 ng/μg
    Biological Activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular Weight
    7.8 kDa
    Reconstitution Instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage Conditions
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient
  • 250 μg
    US
    Log in for availability
    Log in for availability
    Each (250µG)
    Each (250µG) - $1,763.95
    $1,763.95
    Each (250µG)
       $1,763.95
     

    Supplier Number: 250-23-250UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10773-310
    Protein/Peptide Name
    MDC
    Protein Synonyms
    ABCD-1|Macrophage-Derived Chemokine|STCP-1
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    20299 O88430
    Sequence
    GPYGANVEDS ICCQDYIRHP LPSRLVKEFF WTSKSCRKPG VVLITVKNRD ICADPRQVWV KKLLHKLS
    Protease-free
    Y
    Endotoxin Level
    Low
    Endotoxin Content
    >0.1 ng/μg
    Biological Activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular Weight
    7.8 kDa
    Reconstitution Instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage Conditions
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient
  • 500 μg
    US
    Log in for availability
    Log in for availability
    Each (500µG)
    Each (500µG) - $3,016.69
    $3,016.69
    Each (500µG)
       $3,016.69
     

    Supplier Number: 250-23-500UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10773-312
    Protein/Peptide Name
    MDC
    Protein Synonyms
    ABCD-1|Macrophage-Derived Chemokine|STCP-1
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    20299 O88430
    Sequence
    GPYGANVEDS ICCQDYIRHP LPSRLVKEFF WTSKSCRKPG VVLITVKNRD ICADPRQVWV KKLLHKLS
    Protease-free
    Y
    Endotoxin Level
    Low
    Endotoxin Content
    >0.1 ng/μg
    Biological Activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular Weight
    7.8 kDa
    Reconstitution Instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage Conditions
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient
  • 1 mg
    US
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $5,043.82
    $5,043.82
    Each (1mg)
       $5,043.82
     

    Supplier Number: 250-23-1MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10773-314
    Protein/Peptide Name
    MDC
    Protein Synonyms
    ABCD-1|Macrophage-Derived Chemokine|STCP-1
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    20299 O88430
    Sequence
    GPYGANVEDS ICCQDYIRHP LPSRLVKEFF WTSKSCRKPG VVLITVKNRD ICADPRQVWV KKLLHKLS
    Protease-free
    Y
    Endotoxin Level
    Low
    Endotoxin Content
    >0.1 ng/μg
    Biological Activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular Weight
    7.8 kDa
    Reconstitution Instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage Conditions
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient