Mouse Recombinant SDF-1beta (from E. coli)
About this item
SDF-1α and β are stromal-derived, CXC chemokines that signal through the CXCR4 receptor. SDF-1α and β chemoattract B and T cells, and have been shown to induce migration of CD34+ stem cells. Additionally, the SDF-1 proteins exert HIV-suppressive activity in cells expressing the CXCR4 receptor. Human and murine SDF-1 proteins act across species. SDF-1α and β contain the four highly conserved cysteine residues present in CXC chemokines. The mature SDF-1β protein, produced by an N-terminal truncation of two additional amino acids, after removal of the signal sequence, contains 72 amino acid residues. Recombinant Murine SDF-1β (CXCL12) is an 8.5 kDa protein containing 72 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
6 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
SDF-1α and β are stromal-derived, CXC chemokines that signal through the CXCR4 receptor. SDF-1α and β chemoattract B and T cells, and have been shown to induce migration of CD34+ stem cells. Additionally, the SDF-1 proteins exert HIV-suppressive activity in cells expressing the CXCR4 receptor. Human and murine SDF-1 proteins act across species. SDF-1α and β contain the four highly conserved cysteine residues present in CXC chemokines. The mature SDF-1β protein, produced by an N-terminal truncation of two additional amino acids, after removal of the signal sequence, contains 72 amino acid residues. Recombinant Murine SDF-1β (CXCL12) is an 8.5 kDa protein containing 72 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 10773-280
- 10773-282
- 10773-284
- 10773-286
- 10773-288
- 10773-290
- Protein/Peptide Name:
- SDF-1beta
- SDF-1beta
- SDF-1beta
- SDF-1beta
- SDF-1beta
- SDF-1beta
- Protein Synonyms:
- PBSF|Stromal-Cell Derived Factor-1
- PBSF|Stromal-Cell Derived Factor-1
- PBSF|Stromal-Cell Derived Factor-1
- PBSF|Stromal-Cell Derived Factor-1
- PBSF|Stromal-Cell Derived Factor-1
- PBSF|Stromal-Cell Derived Factor-1
- Protein/Peptide Type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Species:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- 20315 P40224
- 20315 P40224
- 20315 P40224
- 20315 P40224
- 20315 P40224
- 20315 P40224
- Sequence:
- KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRL KM
- KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRL KM
- KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRL KM
- KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRL KM
- KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRL KM
- KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRL KM
- Protease-free:
- Y
- Y
- Y
- Y
- Y
- Y
- Endotoxin Level:
- Low
- Low
- Low
- Low
- Low
- Low
- Endotoxin Content:
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- Biological Activity:
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Purity:
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Formulation:
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Molecular Weight:
- 8.5 kDa
- 8.5 kDa
- 8.5 kDa
- 8.5 kDa
- 8.5 kDa
- 8.5 kDa
- Reconstitution Instructions:
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Storage Conditions:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping Temperature:
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
Specifications
Product Family Options
Product Information
- SizeOriginAvailabilityPrice
- Supplier Number: 250-20B-2UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10773-280Protein/Peptide NameSDF-1betaProtein SynonymsPBSF|Stromal-Cell Derived Factor-1Protein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID20315 P40224SequenceKPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRL KMProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight8.5 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: 250-20B-10UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10773-282Protein/Peptide NameSDF-1betaProtein SynonymsPBSF|Stromal-Cell Derived Factor-1Protein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID20315 P40224SequenceKPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRL KMProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight8.5 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: 250-20B-100UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10773-284Protein/Peptide NameSDF-1betaProtein SynonymsPBSF|Stromal-Cell Derived Factor-1Protein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID20315 P40224SequenceKPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRL KMProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight8.5 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: 250-20B-250UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10773-286Protein/Peptide NameSDF-1betaProtein SynonymsPBSF|Stromal-Cell Derived Factor-1Protein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID20315 P40224SequenceKPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRL KMProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight8.5 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: 250-20B-500UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10773-288Protein/Peptide NameSDF-1betaProtein SynonymsPBSF|Stromal-Cell Derived Factor-1Protein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID20315 P40224SequenceKPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRL KMProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight8.5 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - Supplier Number: 250-20B-1MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10773-290Protein/Peptide NameSDF-1betaProtein SynonymsPBSF|Stromal-Cell Derived Factor-1Protein/Peptide TypeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID20315 P40224SequenceKPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRL KMProtease-freeYEndotoxin LevelLowEndotoxin Content>0.1 ng/μgBiological ActivityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular Weight8.5 kDaReconstitution InstructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage ConditionsStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient



