Human;Mouse Recombinant FGF-8b (from E. coli)
Supplier: Peprotech
Certificates
About this item
FGF-8 (FGF-8b) is a heparin-binding growth factor belonging to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. There are 4 known alternate spliced forms of FGF8; FGF-8a, FGF-8b, FGF-8e and FGF-8f. The human and murine FGF-8a and b are identical, unlike human and mouse FGF-8e and f, which are 98% identical. FGF-8 targets mammary carcinoma cells and other cells expressing the FGF receptors. Recombinant Human/Murine FGF-8b is a 22.5 kDa protein consisting of 194 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:HumanMouse
- Storage Conditions:Store at –20 °C prior to reconstitution.
- Endotoxin Content:>0.1 ng/μg
- Biological Activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:2253 P55075
- Reconstitution Instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein Synonyms:Fibroblast Growth Factor-8|AIGF|HBGF-8
- Protein/Peptide Name:FGF-8b
- Purity:0.95
- Molecular Weight:22.5 kDa
- Sequence:MQVTVQSSPN FTQHVREQSL VTDQLSRRLI RTYQLYSRTS GKHVQVLANK RINAMAEDGD PFAKLIVETD TFGSRVRVRG AETGLYICMN KKGKLIAKSN GKGKDCVFTE IVLENNYTAL QNAKYEGWYM AFTRKGRPRK GSKTRQHQRE VHFMKRLPRG HHTTEQSLRF EFLNYPPFTR SLRGSQRTWA PEPR
- Endotoxin Level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together