About this item
IGF-BPs control the distribution, function and activity of IGFs in various cell tissues and body fluids. Currently, there are seven named IGF-BPs that form high affinity complexes with both IGF-I and IGF-II. IGF-BP7 is expressed in a wide range of normal human tissues, and it generally shows reduced expression in cancer cell lines of prostate, breast, colon, and lung origin. It plays a role in skeletal myogenesis by binding to IGF in a manner that inhibits IGF-induced differentiation of skeletal myoblasts, without affecting IGF-induced proliferation. Additionally, IGF-BP7 suppresses growth and colony formation of prostate and breast cancer cell lines through an IGF-independent mechanism, which causes a delay in the G1 phase of the cell cycle and increased apoptosis. Recombinant Human IGF-BP7 is a 26.4 kDa protein consisting of 256 amino acid residues.Manufactured using all Animal-Free reagents.
- Carrier free
- Animal-free
SSSDTCGPCE PASCPPLPPL GCLLGETRDA CGCCPMCARG EGEPCGGGGA GRGYCAPGME CVKSRKRRKG KAGAAAGGPG VSGVCVCKSR YPVCGSDGTT YPSGCQLRAA SQRAESRGEK AITQVSKGTC EQGPSIVTPP KDIWNVTGAQ VYLSCEVIGI PTPVLIWNKV KRGHYGVQRT ELLPGDRDNL AIQTRGGPEK HEVTGWVLVS PLSKEDAGEY ECHASNSQGQ ASASAKITVV DALHEIPVKK GEGAEL
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:−20 °C
- Endotoxin Content:<0.1 ng/μg of protein (<1 EU/μg).
- Biological Activity:Testing In Progress.
- Animal-Free:Y
- Protein/Peptide Name:IGFBP7
- Purity:Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Formulation:Lyophilized
- Shipping Temperature:RT