Mouse Recombinant SCF (from E. coli)
About this item
SCF is a hematopoietic growth factor that exerts its activity by signaling through the c-Kit receptor. SCF and c-Kit are essential for the survival, proliferation and differentiation of hematopoietic cells committed to the melanocyte and germ cell lineages. Human SCF manifests low activity on murine cells, while murine and rat SCF are fully active on human cells. The human SCF gene encodes for a 273 amino acid transmembrane protein, which contains a 25 amino acid N-terminal signal sequence, a 189 amino acid extracellular domain, a 23 amino acid transmembrane domain, and a 36 amino acid cytoplasmic domain. The secreted soluble form of SCF is generated by proteolytic processing of the membrane anchored precursor. Recombinant Murine SCF is an 18.3 kDa polypeptide containing 165 amino acid residues, which corresponds to the sequence of the secreted soluble form of SCF. Manufactured using all Animal-Free reagents.
- Carrier free
- Animal-free
MKEICGNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV IQLSLSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVLC MEENAPKNIK ESPKRPETRS FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
: For research use only. Not for use in diagnostic or therapeutic procedures.
6 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
SCF is a hematopoietic growth factor that exerts its activity by signaling through the c-Kit receptor. SCF and c-Kit are essential for the survival, proliferation and differentiation of hematopoietic cells committed to the melanocyte and germ cell lineages. Human SCF manifests low activity on murine cells, while murine and rat SCF are fully active on human cells. The human SCF gene encodes for a 273 amino acid transmembrane protein, which contains a 25 amino acid N-terminal signal sequence, a 189 amino acid extracellular domain, a 23 amino acid transmembrane domain, and a 36 amino acid cytoplasmic domain. The secreted soluble form of SCF is generated by proteolytic processing of the membrane anchored precursor. Recombinant Murine SCF is an 18.3 kDa polypeptide containing 165 amino acid residues, which corresponds to the sequence of the secreted soluble form of SCF. Manufactured using all Animal-Free reagents.
- Carrier free
- Animal-free
MKEICGNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV IQLSLSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVLC MEENAPKNIK ESPKRPETRS FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 10771-728
- 10771-730
- 10771-732
- 10771-734
- 10771-736
- 10771-738
- Protein/Peptide Name:
- SCF
- SCF
- SCF
- SCF
- SCF
- SCF
- Protein/Peptide Type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Species:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Animal-Free:
- Y
- Y
- Y
- Y
- Y
- Y
- Endotoxin Content:
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- <0.1 ng/μg of protein (<1 EU/μg).
- Biological Activity:
- The ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 10.0 ng/ml, corresponding to a specific activity of ≥ 1 x 105 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 10.0 ng/ml, corresponding to a specific activity of ≥ 1 x 105 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 10.0 ng/ml, corresponding to a specific activity of ≥ 1 x 105 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 10.0 ng/ml, corresponding to a specific activity of ≥ 1 x 105 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 10.0 ng/ml, corresponding to a specific activity of ≥ 1 x 105 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 10.0 ng/ml, corresponding to a specific activity of ≥ 1 x 105 units/mg.
- Purity:
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Greater than 98% by SDS-PAGE gel and HPLC analyses.
- Formulation:
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Storage Conditions:
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- Shipping Temperature:
- RT
- RT
- RT
- RT
- RT
- RT
Specifications
Product Family Options
Product Information
- SizeOriginAvailabilityPrice
- Supplier Number: AF-250-03-10UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10771-728Protein/Peptide NameSCFProtein/Peptide TypeRecombinantSpeciesMouseSourceE. coliAnimal-FreeYEndotoxin Content<0.1 ng/μg of protein (<1 EU/μg).Biological ActivityThe ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 10.0 ng/ml, corresponding to a specific activity of ≥ 1 x 105 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage Conditions−20 °CShipping TemperatureRT - Supplier Number: AF-250-03-50UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10771-730Protein/Peptide NameSCFProtein/Peptide TypeRecombinantSpeciesMouseSourceE. coliAnimal-FreeYEndotoxin Content<0.1 ng/μg of protein (<1 EU/μg).Biological ActivityThe ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 10.0 ng/ml, corresponding to a specific activity of ≥ 1 x 105 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage Conditions−20 °CShipping TemperatureRT - Supplier Number: AF-250-03-100UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10771-732Protein/Peptide NameSCFProtein/Peptide TypeRecombinantSpeciesMouseSourceE. coliAnimal-FreeYEndotoxin Content<0.1 ng/μg of protein (<1 EU/μg).Biological ActivityThe ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 10.0 ng/ml, corresponding to a specific activity of ≥ 1 x 105 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage Conditions−20 °CShipping TemperatureRT - Supplier Number: AF-250-03-250UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10771-734Protein/Peptide NameSCFProtein/Peptide TypeRecombinantSpeciesMouseSourceE. coliAnimal-FreeYEndotoxin Content<0.1 ng/μg of protein (<1 EU/μg).Biological ActivityThe ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 10.0 ng/ml, corresponding to a specific activity of ≥ 1 x 105 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage Conditions−20 °CShipping TemperatureRT - Supplier Number: AF-250-03-500UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10771-736Protein/Peptide NameSCFProtein/Peptide TypeRecombinantSpeciesMouseSourceE. coliAnimal-FreeYEndotoxin Content<0.1 ng/μg of protein (<1 EU/μg).Biological ActivityThe ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 10.0 ng/ml, corresponding to a specific activity of ≥ 1 x 105 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage Conditions−20 °CShipping TemperatureRT - Supplier Number: AF-250-03-1MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10771-738Protein/Peptide NameSCFProtein/Peptide TypeRecombinantSpeciesMouseSourceE. coliAnimal-FreeYEndotoxin Content<0.1 ng/μg of protein (<1 EU/μg).Biological ActivityThe ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 10.0 ng/ml, corresponding to a specific activity of ≥ 1 x 105 units/mg.PurityGreater than 98% by SDS-PAGE gel and HPLC analyses.FormulationLyophilizedStorage Conditions−20 °CShipping TemperatureRT



